3-Amino-1H-indazole

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 1H-Indazol-3-amine, Indazol-3-amine, 1H-Indazol-3-ylamine, Indazol-3-ylamine, 3-Amino-1H-indazole

23261-25G 23261-1G 23261-5G
102563-866EA 305.91 USD
102563-866 102624-592 102624-594
3-Amino-1H-indazole
3-Amino-1H-indazole
Formula: C₇H₇N₃
MW: 133.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00182045
CAS Number: 874-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR