TBS pH 7.5, 10X, Rockland Immunochemicals
Supplier: Rockland Immunochemical
10X Tris buffered saline (TBS) pH 7.5
- Concentrated 10X
- 1.0 M Tris HCl and 1.5 M Sodium Chloride, pH 7.5
- Buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, ELISA, and more.
Tris buffered saline is suitable for multiple applications including biological diluent buffer for antibodies or other biologics. Also may be used as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA. Other applications may require detergents or other additional components.
Application Note: This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. 10X Tris Buffered Saline consists of 1.0 M Tris HCl and 1.5 M Sodium Chloride, pH 7.5, meticulously prepared using ultra pure reagents dissolved in pharmaceutical grade water (WFI).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...