Swine Serum Non-Sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D217-00-1000 D217-00-0100 D217-00-0050 D217-00-0500
75932-514EA 932.99 USD
75932-514 75932-516 75932-518 75932-520
Swine Serum Non-Sterile, Rockland Immunochemicals
Sera (Serum)
Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Swine Serum is used as a component of bioassays, immunoassays or enzyme assays. Swine Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Swine Serum is also available as a sterile preparation for use as a cell culture supplement.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal