Hamster Serum Non-Sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D206-00-0100 D206-00-0050
75928-696EA 1144.03 USD
75928-696 75928-698
Hamster Serum Non-Sterile, Rockland Immunochemicals
Sera (Serum)
Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Hamster Serum is used as a component of bioassays, immunoassays or enzyme assays. Hamster Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Hamster Serum is also available as a sterile preparation for use as a cell culture supplement. Hamster Serum is ideal for investigators in Cancer, Immunology and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal