Rat Recombinant VEGF-165
Supplier: Rockland Immunochemical
012-001-B99-1000 012-001-B99-0010 012-001-B99-0100
75933-494EA 6098 USD
75933-494 75933-498 75933-502 75933-506
Rat Recombinant VEGF-165
Proteins and Peptides
Recombinant Rat VEGF-165 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Vascular Endothelial Growth Factor-A (VEGF-A) was originally isolated from tumor cells and is produced by a wide variety of cell types. In addition to stimulating vascular growth and vascular permeability, VEGF-A may play a role in stimulating vasodilatation via nitric oxide-dependent pathways. VEGF-A has several variants, VEGF-165 being the most abundant. Rat and bovine VEGF are one amino acid shorter than the human factor, and the bovine and human sequences show a homology of 95%. Recombinant rat VEGF-165 is a disulfide linked homodimer, containing two 165 amino acids chains, with a total molecular weight of 38.8 kDa.
Endothelial Growth Factor-165 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Endothelial Growth Factor-165 in immunological assays.