Mouse Recombinant CCL7
Supplier: Rockland Immunochemical
010-001-V76-0100 010-001-V76-0010
75930-050EA 698.96 USD
75930-050 75933-902 75933-908
Mouse Recombinant CCL7
Proteins and Peptides
Recombinant Mouse CCL7 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Monocyte Chemotactic Protein 3 (MCP-3), also called CCL7, is produced by macrophages and some tumor cell lines. MCP-3 signals through three different G protein-coupled receptors, CCR1, CCR2, and CCR3. CCL7 chemoattracts monocytes and can regulate macrophage function. Recombinant mouse MCP-3 is a non-glycosylated protein, containing 74 amino acids, with a molecular weight of 8.5 kDa.
Monocyte Chemotactic Protein-3 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Monocyte Chemotactic Protein-3 in immunological assays.