Human Recombinant IGF-I (from E. coli)
Supplier: Rockland Immunochemical
009-F01-V21-1000 009-F01-V21-0100 009-F01-V21-0020
75928-940EA 518 USD
75928-940 75928-942 75928-944
Human Recombinant IGF-I (from E. coli)
Proteins and Peptides
Recombinant Human IGF-I control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Insulin-like Growth Factor I, IGF-I, is a growth factor produced by the liver when stimulated with growth hormone and can be found circulating throughout the body . IGF-I activates the IGF-I receptor (IGF1R) and the insulin receptor to mediate growth of almost every cell of the body. IGF-I is known as one of the most potent activators of the AKT signaling pathway which is known to be a stimulator of proliferation and an inhibitor of programmed cell death. Mature human IGF-I is 100% homologous with bovine and porcine proteins. Recombinant human IGF-I is a non-glycosylated protein, containing 70 amino acids, with a molecular weight of 7.7 kDa.
Insulin-like Growth Factor I Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Insulin-like Growth Factor I in immunological assays.