Human Recombinant Myostatin Propeptide
Supplier: Rockland Immunochemical
009-001-W02-0100 009-001-W02-0025 009-001-W02-0005
75930-182EA 8502 USD
75930-182 75930-184 75930-186 75930-188
Human Recombinant Myostatin Propeptide
Proteins and Peptides
Recombinant Human Myostatin Propeptide control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Myostatin (GDF-8), a member of the TGF-β superfamily, is a potent and specific negative regulator of skeletal muscle mass. The myostatin propeptide is known to bind and inhibit myostatin in vitro. This interaction is relevant in vivo, with a majority (>70%) of myostatin in serum bound to its propeptide acting as a negative regulator of myostatin. Recombinant human Myostatin Propeptide is a non-glycosylated protein, containing 244 amino acids, with a molecular weight of 27.8 kDa.
Myostatin Propeptide Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Myostatin Propeptide in immunological assays.