Human Recombinant ITAC (CXCL11)
Supplier: Rockland Immunochemical
009-001-V70-0100 009-001-V70-0020 009-001-V70-0005
75929-348EA 4839 USD
75929-348 75929-350 75929-352 75929-354
Human Recombinant ITAC (CXCL11)
Proteins and Peptides
Recombinant Human ITAC (CXCL11) control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interferon Inducible T cell Alpha Chemokine (I-TAC), or CXCL11, expressed at high levels in leukocytes, pancreas and liver exposed to IFNα, β, and γ. I-TAC is one of three chemokines known to bind the receptor CXCR3 (the two others being CXCL9 and CXCL10 (IP-10)) to act as a chemoattractant for IL-2 activated T cells. CXCL11 differs from the other CXCR3 ligands in that it has a higher receptor affinity, thus acts as a stronger agonist. Recombinant human I-TAC is a non-glycosylated protein, containing 73 amino acids, with a molecular weight of 8.3 kDa.
ITAC Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-ITAC in immunological assays.