Human Recombinant Ciliary NTF
Supplier: Rockland Immunochemical
009-001-U71-1000 009-001-U71-0100 009-001-U71-0020 009-001-U71-0005
10802-394EA 4839 USD
10802-394 10802-396 10802-398 10802-400
Human Recombinant Ciliary NTF
Proteins and Peptides
Recombinant Human Ciliary NTF control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Ciliary Neurotrophic Factor (CNTF) is a neurotrophic factor that promotes the survival of various neuronal cell types and may play an important role in the injury response in the nervous system. CNTF, like FGF acidic, FGF basic, and PD-ECGF (platelet-derived endothelial cell growth factor), does not possess a signal sequence that would allow secretion of the factor by classical secretion pathways (endoplasmatic reticulum/Golgi system), but the mechanism underlying the release of CNTF is unknown. Recombinant human CNTF is a non-glycosylated protein, contains 199 amino acids, with a molecular weight of 22.7 kDa.
Ciliary Neurotrophic Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Ciliary Neurotrophic Factor in immunological assays.