(-)-Perillyl alcohol ≥96%

Synonyms: (S)-4-Isopropenyl-1-cyclohexenylmethanol, (S)-p-Mentha-1,8-dien-7-ol, (S)-(4-(Prop-1-en-2-yl)cyclohex-1-en-1-yl)methanol, (S)-(−)-Perillyl alcohol

218391-50G 218391-10G
220043-189EA 202.46 USD
220043-189 220043-188
(-)-Perillyl alcohol ≥96%
(-)-Perillyl alcohol
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
MDL Number: MFCD00062995
CAS Number: 18457-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR