Anti-FCGR2B Rabbit Monoclonal Antibody [clone: 046] PE (Phycoerythrin)

Supplier: Sino Biological
90014-R046-P-25 90014-R046-P-100
103629-550EA 111.1 USD
103629-550 103632-458
Anti-FCGR2B Rabbit Monoclonal Antibody [clone: 046] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus CD32b/FCGR2B/Fc gamma RIIB ( Catalog#90014-C08H; AAL92097.1; Met1-Pro217) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: Anti-CD32B Antibody
Clonality: Monoclonal
Clone: Clone: 046
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR