Anti-CD8A Rabbit Monoclonal Antibody [clone: 082] PE (Phycoerythrin)

Supplier: Sino Biological
60001-R082-P-25 60001-R082-P-100
103630-202EA 111.1 USD
103630-202 103632-382
Anti-CD8A Rabbit Monoclonal Antibody [clone: 082] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Ferret CD8a / Lyt2 (rF CD8a / Lyt2; Catalog#60001-F08H; ABS50091.1; Met1-Glu186) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: Clone: 082
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Ferret
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR