Anti-CD6 Rabbit Monoclonal Antibody [clone:021] PerCP (Peridinin-Chlorophyll Protein Complex)

Supplier: Sino Biological
50711-R021-C-25 50711-R021-C-100
103630-146EA 151.5 USD
103630-146 103632-310
Anti-CD6 Rabbit Monoclonal Antibody [clone:021] PerCP (Peridinin-Chlorophyll Protein Complex)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD6 (rM CD6; Catalog#50711-M08H; Q91WN5; Met1-Gly396) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.

Type: Primary
Antigen: CD6 (PerCP)
Clonality: Monoclonal
Clone: Clone: 021
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR