Anti-EphA2 Rabbit Monoclonal Antibody [clone:101] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
50586-R101-F-25 50586-R101-F-100
103630-108EA 90.9 USD
103630-108 103631-952
Anti-EphA2 Rabbit Monoclonal Antibody [clone:101] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA2 / Eph Receptor A2 (rM EphA2 / Eph Receptor A2; Catalog#50586-M08H; NP_034269.2; Met1-Asn535) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: EphA2
Clonality: Monoclonal
Clone: Clone: 101
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR