Anti-EphA2 Rabbit Monoclonal Antibody [clone:101] APC (Allophycocyanin)

Supplier: Sino Biological
50586-R101-A-25 50586-R101-A-100
103630-106EA 111.1 USD
103630-106 103631-950
Anti-EphA2 Rabbit Monoclonal Antibody [clone:101] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA2/Eph Receptor A2 (rM EphA2/Eph Receptor A2; Catalog#50586-M08H; NP_034269.2; Met1-Asn535) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: EphA2
Clonality: Monoclonal
Clone: Clone: 101
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR