Anti-FCGR3 Rabbit Monoclonal Antibody [clone:002] PE (Phycoerythrin)

Supplier: Sino Biological
50326-R002-P-25 50326-R002-P-100
103630-022EA 30.3 USD
103630-022 103632-198
Anti-FCGR3 Rabbit Monoclonal Antibody [clone:002] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD16 / FCGR3 (rM CD16 / FCGR3; Catalog#50326-M08H; P08508-1; Met1-Thr215) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: Clone: 002
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR