Anti-FCGR1 Rabbit Monoclonal Antibody [clone:027] PE (Phycoerythrin)

Supplier: Sino Biological
50086-R027-P-25 50086-R027-P-100
103629-936EA 111.1 USD
103629-936 103632-132
Anti-FCGR1 Rabbit Monoclonal Antibody [clone:027] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD64 / FCGR1 (rM CD64 / FCGR1; Catalog#50086-M08H; NP_034316.1; Met 1-Pro 297) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: Clone: 027
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR