Anti-CD59 Rabbit Monoclonal Antibody [clone:029] PE (Phycoerythrin)

Supplier: Sino Biological
12474-R029-P-25 12474-R029-P-100
103631-402EA 70.7 USD
103631-402 103632-018
Anti-CD59 Rabbit Monoclonal Antibody [clone:029] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD59 (rh CD59; Catalog#12474-H08H; P13987-1; Met1-Glu101) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: Clone: 029
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR