Anti-CRTAM Mouse Monoclonal Antibody [clone:12] PE (Phycoerythrin)

Supplier: Sino Biological
11975-MM12-P-25 11975-MM12-P-100
103629-698EA 90.9 USD
103629-698 103631-880
Anti-CRTAM Mouse Monoclonal Antibody [clone:12] PE (Phycoerythrin)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CRTAM (rh CRTAM; Catalog#11975-H08H; NP_062550.2; Met1-Ser286) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CRTAM
Clonality: Monoclonal
Clone: Clone: 12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR