Anti-NME1 Mouse Monoclonal Antibody [clone:01] PE (Phycoerythrin)

Supplier: Sino Biological
11615-MM01-P-25 11615-MM01-P-100
103629-606EA 111.1 USD
103629-606 103631-776
Anti-NME1 Mouse Monoclonal Antibody [clone:01] PE (Phycoerythrin)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NME1 / NDKA (rh NME1 / NDKA; Catalog#11615-H07E; NP_000260.1; Ala 2-Glu 152) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: NME1
Clonality: Monoclonal
Clone: Clone: 01
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR