Anti-MST1R Mouse Monoclonal Antibody [clone:03] PE (Phycoerythrin)

Supplier: Sino Biological
11608-MM03-P-25 11608-MM03-P-100
103629-588EA 70.7 USD
103629-588 103631-758
Anti-MST1R Mouse Monoclonal Antibody [clone:03] PE (Phycoerythrin)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD136/MST1R (rh CD136/MST1R; Catalog#11608-H08H; Q04912; Met1-Leu571) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: RON
Clonality: Monoclonal
Clone: Clone: 03
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR