Anti-ALDH1A1 Mouse Monoclonal Antibody [clone:03] PE (Phycoerythrin)

Supplier: Sino Biological
11388-MM03-P-25 11388-MM03-P-100
103629-488EA 111.1 USD
103629-488 103631-634
Anti-ALDH1A1 Mouse Monoclonal Antibody [clone:03] PE (Phycoerythrin)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ALDH1A1 / ALDH1 (rh ALDH1A1 / ALDH1; Catalog#11388-H07E; NP_000680.2; Ser2-Ser501) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: ALDH1A1
Clonality: Monoclonal
Clone: Clone: 03
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR