Anti-IGSF3 Mouse Monoclonal Antibody [clone:05] PE (Phycoerythrin)

Supplier: Sino Biological
11290-MM05-P-25 11290-MM05-P-100
103631-252EA 111.1 USD
103631-252 103631-734
Anti-IGSF3 Mouse Monoclonal Antibody [clone:05] PE (Phycoerythrin)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IGSF3 (rh IGSF3; Catalog#11290-H08H; O75054-1; Met1-Ala1124) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: IGSF3
Clonality: Monoclonal
Clone: Clone: 05
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR