Anti-ADGRE5 Rabbit Monoclonal Antibody [clone:024] PE (Phycoerythrin)

Supplier: Sino Biological
11280-R024-P-25 11280-R024-P-100
103631-382EA 90.9 USD
103631-382 103631-728
Anti-ADGRE5 Rabbit Monoclonal Antibody [clone:024] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD97 (rh CD97; Catalog#11280-H08H; NP_001775.2; Met 1-Gln 398) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD97
Clonality: Monoclonal
Clone: Clone: 024
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR