Anti-CEACAM5 Rabbit Monoclonal Antibody [clone: 061] (PE (Phycoerythrin))

Supplier: Sino Biological
11077-R061-P-25 11077-R061-P-100
103629-414EA 111.1 USD
103629-414 103631-652
Anti-CEACAM5 Rabbit Monoclonal Antibody [clone: 061] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM5 / CD66e (rh CEACAM5 / CD66e; Catalog#11077-H08H; NP_004354.2; Met1-Ala685) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: Clone: 061
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR