Anti-CXADR Rabbit Monoclonal Antibody [clone: 271] (PE (Phycoerythrin))

Supplier: Sino Biological
10799-R271-P-25 10799-R271-P-100
103629-072EA 111.1 USD
103629-072 103630-954
Anti-CXADR Rabbit Monoclonal Antibody [clone: 271] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CXADR / CAR (rh CXADR / CAR; Catalog#10799-H08H; NP_001329.1; Met1-Gly237) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CAR
Clonality: Monoclonal
Clone: Clone: 271
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR