Anti-TEK Rabbit Monoclonal Antibody [clone: 116] PE (Phycoerythrin)

Supplier: Sino Biological
10700-R116-P-100 10700-R116-P-25
103630-898EA 222.2 USD
103630-898 103631-350
Anti-TEK Rabbit Monoclonal Antibody [clone: 116] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human Tie2 / CD202b / TEK (rh Tie2 / CD202b / TEK; Catalog#10700-H08H; AAA61139.1; Met 1-Lys 745) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: TIE2
Clonality: Monoclonal
Clone: Clone: 116
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR