Anti-TEK Rabbit Monoclonal Antibody [clone: 116] APC (Allophycocyanin)

Supplier: Sino Biological
10700-R116-A-25 10700-R116-A-100
103629-170EA 90.9 USD
103629-170 103630-894
Anti-TEK Rabbit Monoclonal Antibody [clone: 116] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human Tie2 / CD202b / TEK (rh Tie2 / CD202b / TEK; Catalog#10700-H08H; AAA61139.1; Met 1-Lys 745) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: TIE2
Clonality: Monoclonal
Clone: Clone: 116
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR