Anti-TEK Mouse Monoclonal Antibody [clone: 07] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
10700-MM07-F-25 10700-MM07-F-100
103629-166EA 70.7 USD
103629-166 103630-890
Anti-TEK Mouse Monoclonal Antibody [clone: 07] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Tie2 / CD202b / TEK (rh Tie2 / CD202b / TEK; Catalog#10700-H08H; AAA61139.1; Met1-Lys745) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: TIE2
Clonality: Monoclonal
Clone: Clone: 07
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR