Anti-TNF Rabbit Monoclonal Antibody [clone: 101] (PE (Phycoerythrin))

Supplier: Sino Biological
10602-R101-P-100 10602-R101-P-25
103630-850EA 222.2 USD
103630-850 103631-330
Anti-TNF Rabbit Monoclonal Antibody [clone: 101] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human TNF-alpha / TNFA (rh TNF-alpha / TNFA; Catalog#10602-HNAE; NP_000585.2; Val77-Leu233) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: Clone: 101
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR