Anti-TNF Rabbit Monoclonal Antibody [clone: 101] (FITC (Fluorescein Isothiocyanate))

Supplier: Sino Biological
10602-R101-F-100 10602-R101-F-25
103630-848EA 151.5 USD
103630-848 103631-328
Anti-TNF Rabbit Monoclonal Antibody [clone: 101] (FITC (Fluorescein Isothiocyanate))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human TNF-alpha (rh TNF-alpha; Catalog#10602-HNAE; NP_000585.2; Val 77-Leu 233) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: Clone: 101
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR