Anti-IL33 Rabbit Monoclonal Antibody [clone: 002] (PE (Phycoerythrin))

Supplier: Sino Biological
10368-R002-P-25 10368-R002-P-100
103628-978EA 111.1 USD
103628-978 103631-560
Anti-IL33 Rabbit Monoclonal Antibody [clone: 002] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-33 (rh IL-33; Catalog#10368-HNAE; NP_254274.1; Ser 112-Thr 270) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: IL-33
Clonality: Monoclonal
Clone: Clone: 002
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR