Anti-IL2RA Rabbit Monoclonal Antibody [clone: 216] (PE (Phycoerythrin))

Supplier: Sino Biological
10165-R216-P-100 10165-R216-P-25
103630-544EA 70.7 USD
103630-544 103631-178
Anti-IL2RA Rabbit Monoclonal Antibody [clone: 216] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD25 / IL2RA (rh CD25 / IL2RA; Catalog#10165-H08H; NP_000408.1; Met1-Cys213) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: Clone: 216
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR