Anti-L1CAM Rabbit Monoclonal Antibody [clone: 014] (PE (Phycoerythrin))

Supplier: Sino Biological
10140-R014-P-25 10140-R014-P-100
103628-808EA 70.7 USD
103628-808 103630-490
Anti-L1CAM Rabbit Monoclonal Antibody [clone: 014] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD171 / N-CAML1 / L1CAM (rh CD171 / N-CAML1 / L1CAM; Catalog#10140-H08H; NP_000416.1; Met 1-Glu 1120) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: Clone: 014
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR