Anti-CD55 Mouse Monoclonal Antibody [clone: 04] (APC (Allophycocyanin))

Supplier: Sino Biological
10101-MM04-A-100 10101-MM04-A-25
103630-436EA 282.8 USD
103630-436 103631-136
Anti-CD55 Mouse Monoclonal Antibody [clone: 04] (APC (Allophycocyanin))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD55/DAF (rh CD55/DAF; Catalog#10101-H08H; NP_000565.1; Met1-Ser353) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: Clone: 04
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR