Anti-CD27 Rabbit Monoclonal Antibody [clone: 023] (APC (Allophycocyanin))

Supplier: Sino Biological
10039-R023-A-25 10039-R023-A-100
103628-670EA 70.7 USD
103628-670 103630-374
Anti-CD27 Rabbit Monoclonal Antibody [clone: 023] (APC (Allophycocyanin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant human CD27 (rh CD27; Catalog#10039-H03H; Met 1-Ile 192; NP_001233.1) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: Clone: 023
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR