Anti-BCL2 Rabbit Monoclonal Antibody [clone: 204] (PE (Phycoerythrin))

Supplier: Sino Biological
100126-R204-P-25 100126-R204-P-100
103630-346EA 90.9 USD
103630-346 103632-420
Anti-BCL2 Rabbit Monoclonal Antibody [clone: 204] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Human BCL2/Bcl-2, and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: Bcl-2
Clonality: Monoclonal
Clone: Clone: 204
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR