Influenza A H4N6 (A/mallard/Ohio/657/2002) Recombinant Neuraminidase/NA (Active) Protein (from HEK293 Cells)
Supplier: Sino Biological
40235-VNAHC-100 40235-VNAHC-1500
103633-212EA 242.4 USD
103633-212 103634-080
Influenza A H4N6 (A/mallard/Ohio/657/2002) Recombinant Neuraminidase/NA (Active) Protein (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the influenza A virus (A/mallard/Ohio/657/2002(H4N6)) neuraminidase (ABI47998.1) (Met1-Lys470) was expressed, the cell lysates are collected, and bio-activity was tested.