Avenanthramide-C methyl ester ≥90%

Supplier: Cayman Chemical Company

10011336-500 10011336-1 10011336-5 10011336-10
76293-076EA 65.81 USD
76293-076 76293-078 76293-080 76293-082
Avenanthramide-C methyl ester ≥90%

  • Inhibitor of NF-κB activation


Inhibitor of NF-κB activation that blocks the phosphorylation of IKK and IκB (IC50 ~ 40 μM); dose dependently inhibits the expression and secretion of IL-6, IL-8, and MCP-1 in human aortic endothelial cells.

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥90%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR