PS-1145 ≥98%

Supplier: Cayman Chemical Company

Synonyms: N-(6-Chloro-9H-pyrido[3,4-b]indol-8-yl)nicotinamide, N-(6-chloro-9H-pyrido[3,4-b]indol-8-yl)-3-pyridinecarboxamide, IKK Inhibitor X

14862-500 14862-1 14862-5
76293-054EA 38.18 USD
76293-054 76293-056 76293-058
PS-1145 ≥98%
PS-1145

  • A potent inhibitor of IKKβ


A potent inhibitor of IKKβ (IC50 = 88-100 nM); used to elucidate roles for NF-κB both in cells and in vivo

Formula: C₁₇H₁₁ClN₄O
MW: 322.8 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18089825
CAS Number: 431898-65-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6445f80bdbd86894bc70053779d41f05973341f1&vid=6445f80bdbd86894bc70053779d41f05973341f1&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=674c4a157ab35636a8e1499a74eed4efd06bb69a&vid=674c4a157ab35636a8e1499a74eed4efd06bb69a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR