LY2409881 ≥98%

Supplier: Cayman Chemical Company
Warning

Synonyms: 2-(5-Chloro-2-((3-(4-methylpiperazin-1-yl)propyl)amino)pyrimidin-4-yl)-N-cyclopropylbenzo[b]thiophene-4-carboxamide trihydrochloride, 2-[5-chloro-2-[[3-(4-methyl-1-piperazinyl)propyl]amino]-4-pyrimidinyl]-N-cyclopropyl-benzo[b]thiophene-4-carboxamide, trihydrochloride

18774-1 18774-5 18774-10 18774-25
76293-046EA 41.12 USD
76293-046 76293-048 76293-050 76293-052
LY2409881 ≥98%
LY2409881

  • A selective IKK2 inhibitor


A selective inhibitor of IKK2 (IC50 = 30 nM); suppresses constitutively activated NF-κB and induces apoptosis in lymphoma cells both in vitro and in preclinical mouse models of B and T cell lymphoma

Formula: C₂₄H₂₉ClN₆OS·₃HCl
Storage Temperature: Freezer
MDL Number: MFCD28900676
CAS Number: 946518-60-1
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=797efc935324a76d8befe959318e7ac70397d221&vid=797efc935324a76d8befe959318e7ac70397d221&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b621d23b74c6db948dc73a152bf95f2852999e19&vid=b621d23b74c6db948dc73a152bf95f2852999e19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR