IKK-16 (hydrochloride) ≥95%

Supplier: Cayman Chemical Company

Synonyms: [4-[[4-(1-benzothiophen-2-yl)pyrimidin-2-yl]amino]phenyl]-(4-pyrrolidin-1-ylpiperidin-1-yl)methanone, hydrochloride, [4-[(4-benzo[b]thien-2-yl-2-pyrimidinyl)amino]phenyl][4-(1-pyrrolidinyl)-1-piperidinyl]-methanone, monohydrochloride, IKK 16 HCl, (4-((4-(Benzo[b]thiophen-2-yl)pyrimidin-2-yl)amino)phenyl)(4-(pyrrolidin-1-yl)piperidin-1-yl)methanone hydrochloride, IKK Inhibitor VII, IKK 16 hydrochloride

13313-5 13313-10 13313-25 13313-50
76293-030EA 119.27 USD
76293-030 76293-032 76293-034 76293-036
IKK-16 (hydrochloride) ≥95%
IKK-16 (hydrochloride)

  • A potent inhibitor of IKKs


A potent inhibitor of IκB kinases (IKKs), displaying IC50 values of 200, 40, and 70 nM for IKKα, IKKβ, and IKK complex, respectively, in cell-free assays; effective in cells and in animals

Formula: C₂₈H₂₉N₅OS·HCl
MW: 520.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09971091
CAS Number: 1186195-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d1271b247cda7a35e9d61d6460b5a40698411bd&vid=6d1271b247cda7a35e9d61d6460b5a40698411bd&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17d6e1eebd75a177738fd27f429c694fec4dc8c8&vid=17d6e1eebd75a177738fd27f429c694fec4dc8c8&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce64830c7bb522747f9cf9e32d64a08cf9220ab1&vid=ce64830c7bb522747f9cf9e32d64a08cf9220ab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥95%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR