IKK Inhibitor VI ≥95%

Supplier: Cayman Chemical Company

17276-500 17276-1 17276-5
76293-024EA 78.15 USD
76293-024 76293-026 76293-028
IKK Inhibitor VI ≥95%
IKK Inhibitor VI

  • An inhibitor of IKK2


A potent, cell-permeable, reversible inhibitor of IKK2 (IC50 = 13 nM); used to evaluate the role of the canonical, IκB-dependent NF-κB signaling pathway in cellular responses

Formula: C₁₂H₁₁N₃O₂S
Storage Temperature: Freezer
CAS Number: 354811-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d191fd46fe4139bf9a640b54c871cb343a0ffcc1&vid=d191fd46fe4139bf9a640b54c871cb343a0ffcc1&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17d6e1eebd75a177738fd27f429c694fec4dc8c8&vid=17d6e1eebd75a177738fd27f429c694fec4dc8c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥95%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR