CAY10657 ≥98%

Supplier: Cayman Chemical Company

Synonyms: 3-[(aminocarbonyl)amino]-5-[4-(4-morpholinylmethyl)phenyl]-2-thiophenecarboxamide

11140-5 11140-10
76292-986EA 200.4 USD
76292-986 76292-988
CAY10657 ≥98%
CAY10657

The NF-κB/Rel family of transcription factors induce and coordinate the expression of pro-inflammatory genes including cytokines, chemokines, interferons, MHC proteins, growth factors, and cell adhesion molecules. Under resting conditions NF-κB dimers are retained in the cytoplasm by a member of the IKB family of inhibitory proteins. Upon activation by cytokines or other stimuli, IKK phosphorylates IKB, initiating a signaling cascade to activate gene transcription. CAY10657 is a thiophenecarboximide derivative proposed to inhibit IKK2. Currently, there are no published reports of its biological activity. However, inhibition of NF-κB activation is likely to be of broad utility in the treatment of inflammatory diseases and cancer.

Formula: C₁₇H₂₀N₄O₃S
MW: 360.4 g/mol
Storage Temperature: Freezer
CAS Number: 494772-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=797efc935324a76d8befe959318e7ac70397d221&vid=797efc935324a76d8befe959318e7ac70397d221&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d49712a091fb7567a8ea045a63f361467db4de6&vid=6d49712a091fb7567a8ea045a63f361467db4de6&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=000911d2630ac779732f6131b13b29fc6fd8557e&vid=000911d2630ac779732f6131b13b29fc6fd8557e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR