CAY10575 ≥95%

Supplier: Cayman Chemical Company

Synonyms: 5-(5,6-dimethoxy-1H-benzimidazol-1-yl)-3-[[4-(methylsulphonyl)phenyl]methoxy]-2-thiophenecarboxamide

10011248-1 10011248-5 10011248-10 10011248-25
76292-968EA 47.11 USD
76292-968 76292-970 76292-972 76292-974
CAY10575 ≥95%
CAY10575

  • An inhibitor of IKK-ε


A benzimidazole analog that inhibits IKK-ε with an IC50 value of ~15.8 µM

Formula: C₂₂H₂₁N₃O₆S₂
MW: 487.6 g/mol
Storage Temperature: Freezer
CAS Number: 916985-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eae283a4a3120270cdd44e8764685176749c31b&vid=3eae283a4a3120270cdd44e8764685176749c31b&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b6230a50d997780c351c4c7ee64c36328b2c8e2&vid=9b6230a50d997780c351c4c7ee64c36328b2c8e2&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=611abb994e0267a7b1b5138b2d829c28db933c2c&vid=611abb994e0267a7b1b5138b2d829c28db933c2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥95%
Stability ≥ 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR