StA-IFN-1 ≥98%

Supplier: Cayman Chemical Company

Synonyms: 4-(1-acetyl-1H-indol-3-yl)-2,4-dihydro-5-methyl-3H-pyrazol-3-one

22402-1 22402-5 22402-10
76292-948EA 68.47 USD
76292-948 76292-950 76292-952
StA-IFN-1 ≥98%
StA-IFN-1

  • An IFN induction pathway inhibitor


An IFN induction pathway inhibitor (IC50 = 4.1 µM in a GFP reporter assay for IFN induction); reduces IFN-β mRNA levels; selectively inhibits the IFN induction pathway but not the IFN signaling pathway

Formula: C₁₄H₁₃N₃O₂
MW: 255.3 g/mol
Storage Temperature: Freezer
CAS Number: 300839-31-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae8fffebff4d4f095d53a6db5045b56971bb98d9&vid=ae8fffebff4d4f095d53a6db5045b56971bb98d9&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e58edbe3a3465ce3c43ae51ba0ffe67f7bd040a0&vid=e58edbe3a3465ce3c43ae51ba0ffe67f7bd040a0&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c7813a254b1dee30c055578f84abe2a13fa27ea&vid=5c7813a254b1dee30c055578f84abe2a13fa27ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR