3'3'-cGAMP (sodium salt) ≥98%
Supplier: Cayman Chemical Company
Synonyms:
Adenosine-Guanosine 3',3'-cyclic monophosphate, 3',3'-Cyclic GMP-AMP, Adenylyl-(3'→5')-3'-guanylic acid, cyclic nucleotide, disodium salt, c-GpAp
17966-100
17966-500
76293-144EA
126.48
USD
76293-144
76293-146
3'3'-cGAMP (sodium salt) ≥98%
3'3'-cGAMP (sodium salt)
- A second messenger and inducer of IFN-β production
A second messenger produced by bacteria that regulates chemotaxis, colonization, and other bacterial functions; binds weakly to STING (Kd = 1.04 µM); induces IFN-β mRNA expression in L929 cells (EC50 = 10.5 nM)
|
Specification Test Results
Formulation | A crystalline solid |
Purity | ≥98% |
Stability | ≥ 2 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...