2'3'-cGAMP (sodium salt) ≥98%

Supplier: Cayman Chemical Company

Synonyms: Guanosine-Adenosine 2',3'-cyclic monophosphate, Adenylyl-(3'→5')-2'-guanylic acid, cyclic nucleotide, disodium salt, 2'-3'-Cyclic GMP-AMP

19887-500 19887-1
76293-140EA 317.21 USD
76293-140 76293-142
2'3'-cGAMP (sodium salt) ≥98%
2'3'-cGAMP (sodium salt)

  • A second messenger and inducer of IFN-β production


A second messenger produced from ATP and GTP by cGMP-AMP synthase (cGAS) in the cytoplasm of mammalian cells in response to the presence of DNA; contains a non-canonical 2’5’-phosphodiester bond; binds tightly to the adapter protein STING (Kd = 3.79 nM) and induces IFN-β mRNA expression (EC50 = 19.4 nM in L929 cells)

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81f090f637703e6eaf9c1e95b3a42c49e4621fc2&vid=81f090f637703e6eaf9c1e95b3a42c49e4621fc2&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a85744dd71af5832d370851b8ad1f914ee9b0be&vid=9a85744dd71af5832d370851b8ad1f914ee9b0be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A solid
Purity ≥98%
Stability ≥ 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR