Cyclic di-IMP (sodium salt) ≥95%
Supplier: Cayman Chemical Company
Synonyms:
c-di-IMP, Cyclic di-inosine monophosphate
22485-500
22485-1
76293-132EA
204.35
USD
76293-132
76293-134
Cyclic di-IMP (sodium salt) ≥95%
Cyclic di-IMP (sodium salt)
- A synthetic second messenger
A synthetic second messenger; has mucosal adjuvant properties in vitro and in vivo by the mucosal route; enriches the population of MHC class I and II, CD80, CD86, CD40, and CD54 positive dendritic cells derived from murine bone marrow; stimulates macrophages at 200 ng/ml; intranasal immunization with β-gal increases IgG titers up to 2-fold and induces a Th1/Th2 response
|
Specification Test Results
Formulation | A crystalline solid |
Purity | ≥95% |
Stability | ≥ 2 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...